Assay Details
| Gene Symbol: Adra1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284380.1 | NP_001271309.1 | 2 - 3 | 213 | 119 | |
| NM_001284381.1 | NP_001271310.1 | 3 - 4 | 363 | 119 | ||
| GenBank mRNA | AK004891.1 | - | 3 - 4 | 320 | 119 | |
| BC018330.1 | - | 2 - 3 | 186 | 119 | ||
| BC037002.1 | - | 2 - 3 | 187 | 119 | ||
| BY107791.1 | - | 2 - 3 | 214 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004891.1, BC018330.1, BC037002.1, NM_001284380.1, NM_001284381.1 transcripts.
Set Membership: |
|

Adra1b
Genomic Map