| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100024656.mm.1 |
NM_008795.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024656.mm.1 |
NM_008795.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024656.mm.1 |
TC0100003069.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100024655.mm.1 |
NM_008795.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024655.mm.1 |
NM_008795.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024655.mm.1 |
TC0100003069.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100024661.mm.1 |
NM_008795.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024661.mm.1 |
NM_008795.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024661.mm.1 |
TC0100003069.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100024658.mm.1 |
NM_008795.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024658.mm.1 |
NM_008795.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024658.mm.1 |
TC0100003069.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100024653.mm.1 |
NM_008795.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024653.mm.1 |
NM_008795.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024653.mm.1 |
TC0100003069.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100024654.mm.1 |
NM_008795.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024654.mm.1 |
NM_008795.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024654.mm.1 |
TC0100003069.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100024660.mm.1 |
NM_008795.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024660.mm.1 |
NM_008795.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100024660.mm.1 |
TC0100003069.mm.1 |
MTA TC |