Assay Details
| Gene Symbol: Cdk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008795.2 | NP_032821.1 | 7 - 8 | 893 | 91 | |
| GenBank mRNA | AK004998.1 | - | 7 - 8 | 837 | 91 | |
| AK171569.1 | - | 7 - 8 | 674 | 91 | ||
| BC119617.1 | - | 8 - 9 | 769 | 40061728 | 91 | |
| BC119618.1 | - | 2 - 3 | 160 | 91 | ||
| X69026.1 | - | 7 - 8 | 910 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk18
Genomic Map