Assay Details
| Gene Symbol: Cp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276248.1 | NP_001263177.1 | 14 - 15 | 2771 | 84 | |
| NM_001276250.1 | NP_001263179.1 | 14 - 15 | 2771 | 84 | ||
| NM_007752.3 | NP_031778.2 | 14 - 15 | 2774 | 84 | ||
| GenBank mRNA | AK014835.1 | - | 14 - 15 | 2757 | 84 | |
| AK043248.1 | - | 14 - 15 | 2716 | 84 | ||
| AK080701.1 | - | 6 - 7 | 870 | 84 | ||
| AK160035.1 | - | 10 - 11 | 1700 | 84 | ||
| BC062957.1 | - | 14 - 15 | 2774 | 30547760 | 84 | |
| DQ336396.1 | - | 14 - 15 | 2772 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cp
Genomic Map