Assay Details
| Gene Symbol: Cp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276248.1 | NP_001263177.1 | 16 - 17 | 3086 | 72 | |
| NM_001276250.1 | NP_001263179.1 | 16 - 17 | 3086 | 72 | ||
| NM_007752.3 | NP_031778.2 | 16 - 17 | 3089 | 72 | ||
| GenBank mRNA | AK043248.1 | - | 16 - 17 | 3031 | 72 | |
| AK080701.1 | - | 8 - 9 | 1185 | 72 | ||
| AK160035.1 | - | 12 - 13 | 2015 | 72 | ||
| BC062957.1 | - | 16 - 17 | 3089 | 30547760 | 72 | |
| DQ336396.1 | - | 16 - 17 | 3087 | 72 | ||
| U49430.1 | - | 16 - 17 | 2967 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cp
Genomic Map