Assay Details
| Gene Symbol: Cp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276248.1 | NP_001263177.1 | 3 - 4 | 835 | 119 | |
| NM_001276250.1 | NP_001263179.1 | 3 - 4 | 835 | 119 | ||
| NM_007752.3 | NP_031778.2 | 3 - 4 | 835 | 119 | ||
| GenBank mRNA | AK014835.1 | - | 3 - 4 | 821 | 119 | |
| AK033842.1 | - | 2 - 3 | 244 | 119 | ||
| AK043248.1 | - | 3 - 4 | 780 | 119 | ||
| AK149523.1 | - | 3 - 4 | 671 | 119 | ||
| BC062957.1 | - | 3 - 4 | 835 | 30547760 | 119 | |
| DQ336396.1 | - | 3 - 4 | 833 | 119 | ||
| U49430.1 | - | 3 - 4 | 710 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cp
Genomic Map