Assay Details
| Gene Symbol: Cp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276248.1 | NP_001263177.1 | 4 - 5 | 1005 | 70 | |
| NM_001276250.1 | NP_001263179.1 | 4 - 5 | 1005 | 70 | ||
| NM_007752.3 | NP_031778.2 | 4 - 5 | 1005 | 70 | ||
| GenBank mRNA | AK014835.1 | - | 4 - 5 | 991 | 70 | |
| AK043248.1 | - | 4 - 5 | 950 | 70 | ||
| BC062957.1 | - | 4 - 5 | 1005 | 30547760 | 70 | |
| DQ336396.1 | - | 4 - 5 | 1003 | 70 | ||
| U49430.1 | - | 4 - 5 | 880 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cp
Genomic Map