Assay Details
| Gene Symbol: Cp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276248.1 | NP_001263177.1 | 5 - 6 | 1267 | 69 | |
| NM_001276250.1 | NP_001263179.1 | 5 - 6 | 1267 | 69 | ||
| NM_007752.3 | NP_031778.2 | 5 - 6 | 1267 | 69 | ||
| GenBank mRNA | AK014835.1 | - | 5 - 6 | 1253 | 69 | |
| AK043248.1 | - | 5 - 6 | 1212 | 69 | ||
| AK160035.1 | - | 1 - 2 | 193 | 69 | ||
| BC062957.1 | - | 5 - 6 | 1267 | 30547760 | 69 | |
| DQ336396.1 | - | 5 - 6 | 1265 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cp
Genomic Map