Assay Details
| Gene Symbol: Gyg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013755.3 | NP_038783.1 | 1 - 2 | 121 | 66 | |
| GenBank mRNA | AA553085.1 | - | 1 - 2 | 120 | 66 | |
| AF114031.1 | - | 1 - 2 | 56 | 66 | ||
| AK159576.1 | - | 1 - 2 | 102 | 66 | ||
| AK209105.1 | - | 1 - 2 | 38 | 66 | ||
| BC029903.1 | - | 1 - 2 | 82 | 4487769 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF114031.1, AK159576.1, BC029903.1, NM_013755.3 transcripts.
Set Membership: |
|

Gyg
Genomic Map