Assay Details
| Gene Symbol: Gyg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013755.3 | NP_038783.1 | 3 - 4 | 431 | 85 | |
| GenBank mRNA | AA553085.1 | - | 3 - 4 | 430 | 85 | |
| AF114031.1 | - | 3 - 4 | 366 | 85 | ||
| AK159576.1 | - | 3 - 4 | 412 | 85 | ||
| AK212191.1 | - | 1 - 2 | 149 | 85 | ||
| BC029903.1 | - | 3 - 4 | 392 | 4487769 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gyg
Genomic Map