Assay Details
| Gene Symbol: Nceh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178772.3 | NP_848887.1 | 2 - 3 | 438 | 121 | |
| GenBank mRNA | AK038691.1 | - | 439 | 121 | ||
| AK045363.1 | - | 2 - 3 | 439 | 121 | ||
| AK085767.1 | - | 2 - 3 | 439 | 121 | ||
| AK128968.1 | - | 2 - 3 | 449 | 121 | ||
| AK135837.1 | - | 2 - 3 | 428 | 121 | ||
| AK173158.1 | - | 2 - 3 | 408 | 121 | ||
| BC082569.1 | - | 2 - 3 | 421 | 30550100 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nceh1
Genomic Map