Assay Details
| Gene Symbol: Nhlrc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172501.2 | NP_766089.1 | 2 - 3 | 352 | 109 | |
| GenBank mRNA | AK029465.1 | - | 2 - 3 | 347 | 109 | |
| AK033167.1 | - | 2 - 3 | 352 | 109 | ||
| AK133986.1 | - | 302 | 109 | |||
| BC087903.1 | - | 2 - 3 | 339 | 30250042 | 109 | |
| BC107356.1 | - | 2 - 3 | 340 | 40055875 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nhlrc3
Genomic Map