Assay Details
| Gene Symbol: Nhlrc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172501.2 | NP_766089.1 | 6 - 7 | 907 | 78 | |
| GenBank mRNA | AK029465.1 | - | 6 - 7 | 902 | 78 | |
| AK033167.1 | - | 6 - 7 | 907 | 78 | ||
| AK133986.1 | - | 857 | 78 | |||
| AK220477.1 | - | 824 | 78 | |||
| BC087903.1 | - | 6 - 7 | 894 | 30250042 | 78 | |
| BC107356.1 | - | 6 - 7 | 895 | 40055875 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141943 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141942 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141941 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141946 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141944 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141945 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141947 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641922 | BC107356.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641922 | AK133986.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641922 | AK220477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641922 | BC087903.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641922 | AK029465.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641922 | BC107356.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641922 | AK133986.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641922 | AK220477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641922 | BC087903.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P641922 | AK029465.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383482 | AK039550.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270969 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 725892 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 804155 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904269 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262387 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32594 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554853 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20039 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | AK004110.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | AK122523.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | AY739114.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | AY739115.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | AY739116.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | AY739117.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | AY739118.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | AY739119.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | AY739120.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | AY739121.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | AY739122.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | NM_001008791.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | NM_001008792.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | NM_001008793.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | NM_001008794.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | NM_001008795.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | NM_001008796.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | NM_001008797.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | NM_001008798.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724559 | NM_028640.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 269587 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 72242 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 114731 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10074 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45723 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19242 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136810 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119442 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150155 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180433 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641922 | BC107356.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641922 | AK133986.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641922 | AK220477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641922 | BC087903.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641922 | AK029465.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641922 | BC107356.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641922 | AK133986.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641922 | AK220477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641922 | BC087903.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P641922 | AK029465.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22141921 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136810 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180433 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150155 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119442 | - | - |
| Whole Mouse Genome G4122A | A_52_P320202 | AK220477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P320202 | AK077661.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641922 | BC107356.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641922 | AK133986.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641922 | AK220477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641922 | BC087903.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641922 | AK029465.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641922 | BC107356.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641922 | AK133986.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641922 | AK220477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641922 | BC087903.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P641922 | AK029465.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139291 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150155 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119442 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180433 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136810 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02582230 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02582233 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141945 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141947 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141942 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141944 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141941 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141943 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22141946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22141869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03005511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02877232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02377070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03005510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03005506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02877230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02983394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02582236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02552193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02877240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02304258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02582234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02582235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02582233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22141906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02983397 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10498153 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498149 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498147 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498150 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498154 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498152 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498151 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10498148 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456026_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10498146 | - | - |
| Mouse Genome 430 2.0 Array | 1446204_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456026_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1446204_at | - | - |
| Mouse Genome 430 2.0 Array | 1456026_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300015036.mm.1 | NM_172501.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300015036.mm.1 | NM_172501.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300015036.mm.1 | TC0300001982.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300015035.mm.1 | NM_172501.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300015035.mm.1 | NM_172501.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300015035.mm.1 | TC0300001982.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456026_PM_at | - | - |
| Mouse Expression Set 430 | 1456026_at | - | - |
| Murine Genome U74v2 Set | 166308_PM_at | - | - |
| Murine Genome U74v2 Set | 166308_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456026_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456026_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101770056 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5720520 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5720520 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101770056 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720520 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219639 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219639 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2438516 | - | - |
| mouse-6 v1.1 (Illumina) | scl22162.7.1_2-S | - | - |
| mouse-6 v1.1 (Illumina) | scl22163.1_255-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5720520 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101770056 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22162.7.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720520 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101770056 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1219639 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219639 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2438516 | - | - |

Nhlrc3
Genomic Map