Assay Details
| Gene Symbol: Colec11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027866.1 | NP_082142.1 | 1 - 2 | 61 | 70 | |
| GenBank mRNA | AK003121.1 | - | 1 - 2 | 61 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003121.1, NM_027866.1 transcripts.
Set Membership: |
|

Colec11
Genomic Map