Assay Details
| Gene Symbol: Colec11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027866.1 | NP_082142.1 | 4 - 5 | 354 | 69 | |
| GenBank mRNA | AK003121.1 | - | 4 - 5 | 354 | 69 | |
| BC104380.1 | - | 4 - 5 | 325 | 40050136 | 69 | |
| BC104381.1 | - | 4 - 5 | 325 | 40050143 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Colec11
Genomic Map