Assay Details
| Gene Symbol: Leap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153069.3 | NP_694709.1 | 2 - 3 | 229 | 68 | |
| GenBank mRNA | AJ409055.1 | - | 2 - 3 | 229 | 68 | |
| BC089593.1 | - | 2 - 3 | 222 | 6749700 | 68 | |
| W82161.1 | - | 2 - 3 | 263 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P117881 | BC089593.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P117881 | W82161.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P117881 | AJ409055.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P117881 | NM_153069.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P117881 | AA571276.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P117881 | BC089593.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P117881 | W82161.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P117881 | AJ409055.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P117881 | NM_153069.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P117881 | AA571276.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881515 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173227 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P117881 | BC089593.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P117881 | W82161.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P117881 | AJ409055.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P117881 | NM_153069.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P117881 | AA571276.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P117881 | BC089593.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P117881 | W82161.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P117881 | AJ409055.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P117881 | NM_153069.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P117881 | AA571276.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27804668 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173227 | - | - |
| Whole Mouse Genome G4122A | A_51_P117881 | BC089593.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P117881 | W82161.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P117881 | AJ409055.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P117881 | NM_153069.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P117881 | AA571276.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P117881 | BC089593.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P117881 | W82161.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P117881 | AJ409055.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P117881 | NM_153069.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P117881 | AA571276.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132924 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173227 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09406593 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09406595 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09406601 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09575445 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09406591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27804716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09575441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09192684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09575448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09025423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09406593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09025424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09334426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09025422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09126304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09334425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27804700 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10385815 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385817 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385816 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427480_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10385814 | - | - |
| Mouse Genome 430 2.0 Array | 1427480_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427480_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 259301_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 259301_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100024800.mm.1 | NM_153069.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024800.mm.1 | NM_153069.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024800.mm.1 | TC1100002728.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427480_at | - | - |
| Mouse Expression Set 430 | 1427480_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427480_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427480_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427480_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427480_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 508976 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2470044 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2470044 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002470044 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2649275 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2649275 | - | - |
| mouse-6 v1.1 (Illumina) | scl40238.3.1_75-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2470044 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40238.3.1_75-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2470044 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2649275 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2649275 | - | - |

Leap2
Genomic Map