Assay Details
| Gene Symbol: Fstl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177059.3 | NP_796033.2 | 4 - 5 | 595 | 95 | |
| GenBank mRNA | AF374459.1 | - | 4 - 5 | 595 | 95 | |
| BC132353.1 | - | 4 - 5 | 572 | 40130630 | 95 | |
| BC144824.1 | - | 4 - 5 | 572 | 95 | ||
More Information
Set Membership: |
|

Fstl4
Genomic Map