Assay Details
| Gene Symbol: Cenph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021886.1 | NP_068686.1 | 1 - 2 | 209 | 211 | |
| GenBank mRNA | AB017634.1 | - | 1 - 2 | 209 | 211 | |
| AK006208.1 | - | 1 - 2 | 194 | 210 | ||
| AK011264.1 | - | 1 - 2 | 194 | 211 | ||
| AK011738.1 | - | 1 - 2 | 194 | 211 | ||
| BC025084.1 | - | 1 - 2 | 160 | 3501244 | 211 | |
More Information
Set Membership: |
|

Cenph
Genomic Map