Assay Details
| Gene Symbol: Homer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284189.1 | NP_001271118.1 | 5 - 6 | 1331 | 65 | |
| NM_152134.3 | NP_687036.1 | 5 - 6 | 850 | 65 | ||
| GenBank mRNA | AF093258.1 | - | 5 - 6 | 680 | 65 | |
| AY137385.1 | - | 5 - 6 | 850 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Homer1
Genomic Map