Assay Details
| Gene Symbol: Csf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009969.4 | NP_034099.2 | 2 - 3 | 485 | 125 | |
| GenBank mRNA | AK053196.1 | - | 2 - 3 | 485 | 125 | |
| BC116878.1 | - | 2 - 3 | 219 | 40126197 | 125 | |
| BC116880.1 | - | 2 - 3 | 219 | 40126199 | 125 | |
| EU366957.1 | - | 2 - 3 | 196 | 125 | ||
| M11848.1 | - | 2 - 3 | 154 | 125 | ||
| X02333.1 | - | 2 - 3 | 369 | 125 | ||
| X03019.1 | - | 2 - 3 | 228 | 125 | ||
| X03221.1 | - | 2 - 3 | 227 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Csf2
Genomic Map