Assay Details
| Gene Symbol: Sept8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252332.1 | NP_001239261.1 | 5 - 6 | 975 | 56 | |
| NM_001252333.1 | NP_001239262.1 | 5 - 6 | 969 | 56 | ||
| NM_033144.2 | NP_149156.1 | 5 - 6 | 975 | 56 | ||
| GenBank mRNA | AB093215.1 | - | 5 - 6 | 825 | 56 | |
| AF179996.1 | - | 4 - 5 | 671 | 56 | ||
| AK144178.1 | - | 5 - 6 | 811 | 56 | ||
| BC037707.1 | - | 5 - 6 | 763 | 56 | ||
| BC049819.1 | - | 5 - 6 | 790 | 56 | ||
| BC059248.1 | - | 5 - 6 | 911 | 6400429 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sept8
Genomic Map