Assay Details
| Gene Symbol: Cadm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053199.3 | NP_444429.1 | 2 - 3 | 424 | 122 | |
| GenBank mRNA | AF195662.1 | - | 2 - 3 | 368 | 122 | |
| AK036352.1 | - | 3 - 4 | 480 | 122 | ||
| AK038917.1 | - | 2 - 3 | 381 | 122 | ||
| AK053077.1 | - | 2 - 3 | 378 | 122 | ||
| AY059393.1 | - | 2 - 3 | 223 | 122 | ||
| BC079837.1 | - | 3 - 4 | 464 | 122 | ||
| BC115417.2 | - | 2 - 3 | 329 | 40093410 | 122 | |
| BC115418.2 | - | 2 - 3 | 329 | 40093417 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cadm3
Genomic Map