Assay Details
| Gene Symbol: Usf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK142437.1 | - | 4 - 5 | 376 | 66 | |
| BC049784.1 | - | 4 - 5 | 319 | 6506829 | 66 | |
| CJ050979.1 | - | 4 - 5 | 455 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827837 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827835 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827838 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827834 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827832 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827836 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827839 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169645 | AK142437.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169645 | BC049784.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169645 | AF479773.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169645 | NM_009480.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786660 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29007 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 388454 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538863 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163202 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169645 | AK142437.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169645 | BC049784.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169645 | AF479773.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169645 | NM_009480.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P233478 | BC049784.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827828 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163202 | - | - |
| Whole Mouse Genome G4122A | A_51_P233478 | BC049784.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125537 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120433 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123669 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127737 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163202 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01069568 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01069564 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01069561 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01069574 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00721611 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827836 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827839 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827838 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827837 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827835 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827834 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01069568 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00342635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01069558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01069555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00903398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01074057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00903403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00943058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00938167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01074056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00903405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00464315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00796248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827850 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351620 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351621 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351617 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351618 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351622 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351619 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448805_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426164_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459758_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351616 | - | - |
| Mouse Genome 430 2.0 Array | 1448805_at | - | - |
| Mouse Genome 430 2.0 Array | 1426164_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1459758_at | - | - |
| Mouse Genome 430 2.0 Array | 1459758_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448805_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426164_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22278_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22278_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100013122.mm.1 | NM_009480.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013122.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013123.mm.1 | NM_009480.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013123.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013128.mm.1 | NM_009480.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013128.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013110.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013114.mm.1 | NM_009480.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013114.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013131.mm.1 | NM_009480.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013131.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013133.mm.1 | NM_009480.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013133.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013130.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013113.mm.1 | NM_009480.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013113.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013118.mm.1 | NM_009480.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013118.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013119.mm.1 | NM_009480.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013119.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013121.mm.1 | NM_009480.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013121.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013127.mm.1 | NM_009480.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013127.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013120.mm.1 | NM_009480.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013120.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013129.mm.1 | TC0100003870.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100013132.mm.1 | NM_009480.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100013132.mm.1 | TC0100003870.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426164_a_at | - | - |
| Mouse Expression Set 430 | 1448805_at | - | - |
| Mouse Expression Set 430 | 1448805_PM_at | - | - |
| Mouse Expression Set 430 | 1426164_PM_a_at | - | - |
| Mouse Expression Set 430 | 1459758_at | - | - |
| Mouse Expression Set 430 | 1459758_PM_at | - | - |
| Murine Genome U74v2 Set | 93655_PM_at | - | - |
| Murine Genome U74v2 Set | 93655_at | - | - |
| Murine Genome U74v2 Set | 93656_g_at | - | - |
| Murine Genome U74v2 Set | 93656_PM_g_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426164_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426164_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448805_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448805_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426164_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426164_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448805_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448805_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459758_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459758_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 349549 | - | - |
| ABI Mouse Genome Survey Microarray | 917201 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4280156 | - | - |
| MouseWG-6 v2 Expression BeadChip | 106940471 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6370113 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106940471 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4280156 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6370113 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4610114 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004610114 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004280156 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006370113 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1254639 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2518938 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2435505 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2435505 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2518938 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2436330 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254639 | - | - |
| mouse-6 v1.1 (Illumina) | scl0022278.1_0-S | - | - |
| mouse-6 v1.1 (Illumina) | scl022278.9_243-S | - | - |
| mouse-6 v1.1 (Illumina) | scl022278.1_12-S | - | - |
| mouse-6 v1.1 (Illumina) | scl17225.9_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4280156 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6370113 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106940471 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4610114 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl022278.9_243-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0022278.1_0-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl022278.1_12-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106940471 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4610114 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370113 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280156 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2435505 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1254639 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2435505 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2436330 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2518938 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254639 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl022278.9_243-S | - | - |

Usf1
Genomic Map