Assay Details
| Gene Symbol: Rgs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026380.3 | NP_080656.2 | 3 - 4 | 525 | 60 | |
| GenBank mRNA | AK044337.1 | - | 3 - 4 | 399 | 60 | |
| AK142301.1 | - | 3 - 4 | 394 | 60 | ||
| BC065056.1 | - | 3 - 4 | 458 | 5705033 | 60 | |
| BC094577.1 | - | 2 - 3 | 141 | 6852696 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK044337.1, AK142301.1, BC065056.1, BC094577.1, NM_026380.3 transcripts.
Set Membership: |
|

Rgs8
Genomic Map