Assay Details
| Gene Symbol: Lrrfip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111311.1 | NP_001104781.1 | 2 - 3 | 367 | 83 | |
| NM_001111312.1 | NP_001104782.1 | 2 - 3 | 254 | 83 | ||
| GenBank mRNA | AK041162.1 | - | 2 - 3 | 205 | 83 | |
| AK077637.1 | - | 2 - 3 | 372 | 83 | ||
| AK133661.1 | - | 2 - 3 | 347 | 83 | ||
| AK134120.1 | - | 2 - 3 | 359 | 83 | ||
| BC144955.1 | - | 2 - 3 | 220 | 83 | ||
| BY739242.1 | - | 2 - 3 | 303 | 83 | ||
More Information
Set Membership: |
|

Lrrfip1
Genomic Map