Assay Details
| Gene Symbol: Ackr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271607.1 | NP_001258536.1 | 2 - 3 | 393 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271607.1 transcripts.
Set Membership: |
|

Ackr3
Genomic Map