Assay Details
| Gene Symbol: Nit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242580.1 | NP_001229509.1 | 1 - 2 | 117 | 74 | |
| GenBank mRNA | AK002269.1 | - | 1 - 2 | 124 | 74 | |
| AK004988.2 | - | 78 | 74 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001242580.1 transcripts.
Set Membership: |
|

Nit1
Genomic Map