Assay Details
| Gene Symbol: Nit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242580.1 | NP_001229509.1 | 4 - 5 | 551 | 84 | |
| NM_012049.2 | NP_036179.1 | 4 - 5 | 519 | 84 | ||
| GenBank mRNA | AF069988.1 | - | 4 - 5 | 500 | 84 | |
| AK002269.1 | - | 4 - 5 | 559 | 84 | ||
| AK004988.2 | - | 512 | 84 | |||
| BC021634.1 | - | 4 - 5 | 501 | 4008543 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nit1
Genomic Map