Assay Details
| Gene Symbol: Apoa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BY780589.1 | - | 1 - 2 | 74 | 61 | |
| CB947382.1 | - | 1 - 2 | 41 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apoa2| Gene Symbol: Apoa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BY780589.1 | - | 1 - 2 | 74 | 61 | |
| CB947382.1 | - | 1 - 2 | 41 | 61 | ||
Set Membership: |
|