Assay Details
| Gene Symbol: Apoa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013474.2 | NP_038502.2 | 3 - 4 | 287 | 61 | |
| GenBank mRNA | AK145823.1 | - | 3 - 4 | 250 | 61 | |
| BU609941.1 | - | 438 | 61 | |||
| BY780589.1 | - | 3 - 4 | 285 | 61 | ||
| CB947382.1 | - | 3 - 4 | 252 | 61 | ||
| M79361.1 | - | 3 - 4 | 252 | 61 | ||
| X62772.1 | - | 3 - 4 | 235 | 61 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of cytokine secretion involved in immune response
protein folding
fatty acid metabolic process
phosphatidylcholine biosynthetic process
transport
lipid transport
cholesterol metabolic process
phospholipid catabolic process
response to glucose
positive regulation of cholesterol esterification
negative regulation of very-low-density lipoprotein particle remodeling
beta-glucoside transport
protein oxidation
peptidyl-methionine modification
regulation of intestinal cholesterol absorption
cholesterol transport
organ regeneration
regulation of protein stability
negative regulation of cholesterol transport
cholesterol efflux
phospholipid efflux
triglyceride-rich lipoprotein particle remodeling
low-density lipoprotein particle remodeling
high-density lipoprotein particle remodeling
high-density lipoprotein particle assembly
high-density lipoprotein particle clearance
lipoprotein metabolic process
cholesterol homeostasis
positive regulation of catalytic activity
reverse cholesterol transport
positive regulation of interleukin-8 biosynthetic process
diacylglycerol catabolic process
negative regulation of lipid catabolic process
positive regulation of lipid catabolic process
negative regulation of lipase activity
negative regulation of cholesterol import
negative regulation of cholesterol transporter activity
lipid transporter activity
phospholipid binding
high-density lipoprotein particle binding
lipid binding
cholesterol binding
cholesterol transporter activity
phosphatidylcholine binding
apolipoprotein receptor binding
protein homodimerization activity
protein heterodimerization activity
lipase inhibitor activity
phosphatidylcholine-sterol O-acyltransferase activator activity
high-density lipoprotein particle receptor binding
negative regulation of cytokine secretion involved in immune response
protein folding
fatty acid metabolic process
phosphatidylcholine biosynthetic process
transport
lipid transport
cholesterol metabolic process
phospholipid catabolic process
response to glucose
positive regulation of cholesterol esterification
negative regulation of very-low-density lipoprotein particle remodeling
beta-glucoside transport
protein oxidation
peptidyl-methionine modification
regulation of intestinal cholesterol absorption
cholesterol transport
organ regeneration
regulation of protein stability
negative regulation of cholesterol transport
cholesterol efflux
phospholipid efflux
triglyceride-rich lipoprotein particle remodeling
low-density lipoprotein particle remodeling
high-density lipoprotein particle remodeling
high-density lipoprotein particle assembly
high-density lipoprotein particle clearance
lipoprotein metabolic process
cholesterol homeostasis
positive regulation of catalytic activity
reverse cholesterol transport
positive regulation of interleukin-8 biosynthetic process
diacylglycerol catabolic process
negative regulation of lipid catabolic process
positive regulation of lipid catabolic process
negative regulation of lipase activity
negative regulation of cholesterol import
negative regulation of cholesterol transporter activity
lipid transporter activity
phospholipid binding
high-density lipoprotein particle binding
lipid binding
cholesterol binding
cholesterol transporter activity
phosphatidylcholine binding
apolipoprotein receptor binding
protein homodimerization activity
protein heterodimerization activity
lipase inhibitor activity
phosphatidylcholine-sterol O-acyltransferase activator activity
high-density lipoprotein particle receptor binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966035 | AK145823.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966035 | NM_013474.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966035 | CB947382.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966035 | M79361.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966035 | X62772.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178553 | AK145823.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178553 | BC031786.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178553 | NM_013474.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178553 | CB947382.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2178553 | X62772.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340899 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 94021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358065 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 379635 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52467 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 64590 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139659 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 75777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35555 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174692 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178553 | AK145823.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178553 | BC031786.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178553 | NM_013474.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178553 | CB947382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2178553 | X62772.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966035 | AK145823.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966035 | NM_013474.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966035 | CB947382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966035 | M79361.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966035 | X62772.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128973 | BC031786.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128973 | M79362.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128973 | S79794.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128973 | X04119.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20826753 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174692 | - | - |
| Whole Mouse Genome G4122A | A_51_P128973 | BC031786.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128973 | M79362.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128973 | S79794.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128973 | X04119.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116139 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174692 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00998721 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00998718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01055832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826741 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826737 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00914434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00998736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826744 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351550 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351548 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351547 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351549 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417950_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351546 | - | - |
| Mouse Genome 430 2.0 Array | 1417950_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417950_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11807_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11807_PM_at | - | - |
| Mouse Expression Set 430 | 1417950_a_at | - | - |
| Mouse Expression Set 430 | 1417950_PM_a_at | - | - |
| Murine Genome U74v2 Set | 161924_PM_f_at | - | - |
| Murine Genome U74v2 Set | 99648_at | - | - |
| Murine Genome U74v2 Set | 161924_f_at | - | - |
| Murine Genome U74v2 Set | 99648_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417950_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417950_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417950_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417950_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6510364 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6860411 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6860411 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6510364 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006860411 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006510364 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247156 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2775957 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247156 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2775957 | - | - |
| mouse-6 v1.1 (Illumina) | scl000890.1_7-S | - | - |
| mouse-6 v1.1 (Illumina) | scl011807.2_4-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6510364 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6860411 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000890.1_7-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl011807.2_4-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6860411 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6510364 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1247156 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247156 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2775957 | - | - |

Apoa2
Genomic Map