Assay Details
| Gene Symbol: Cfap126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081275.1 | NP_001074744.1 | 5 - 6 | 628 | 90 | |
| GenBank mRNA | AK005804.1 | - | 5 - 6 | 630 | 90 | |
| BC061017.1 | - | 4 - 5 | 354 | 6774925 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20826100 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20826099 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20826098 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430259 | AK005804.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430259 | BC061017.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430259 | NM_001081275.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430259 | AK005804.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430259 | BC061017.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P430259 | NM_001081275.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2186668 | AK005804.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2186668 | BC061017.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2186668 | NM_001081275.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 834582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45119 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150158 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186668 | AK005804.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186668 | BC061017.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2186668 | NM_001081275.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430259 | AK005804.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430259 | BC061017.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430259 | NM_001081275.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430259 | AK005804.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430259 | BC061017.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P430259 | NM_001081275.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150158 | - | - |
| Whole Mouse Genome G4122A | A_51_P430259 | AK005804.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430259 | BC061017.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430259 | NM_001081275.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430259 | AK005804.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430259 | BC061017.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P430259 | NM_001081275.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150158 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20826099 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20826100 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20826098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20826227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00519957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00942894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00962058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01081251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00062439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00942891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00268040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826225 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01086136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20826217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00306852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00286343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00743835 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351518 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351520 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351522 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351524 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351519 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429181_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351517 | - | - |
| Mouse Genome 430 2.0 Array | 1429181_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429181_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75472_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75472_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100012901.mm.1 | NM_001081275.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012901.mm.1 | NM_001081275.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012901.mm.1 | TC0100001577.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100012899.mm.1 | NM_001081275.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012899.mm.1 | NM_001081275.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012899.mm.1 | TC0100001577.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100012898.mm.1 | NM_001081275.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012898.mm.1 | NM_001081275.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100012898.mm.1 | TC0100001577.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429181_PM_at | - | - |
| Mouse Expression Set 430 | 1429181_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429181_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429181_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 410126 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2645509 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2645509 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2645509 | - | - |

Cfap126
Genomic Map