Assay Details
| Gene Symbol: Lims1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026148.3 | NP_080424.2 | 1 - 2 | 223 | 89 | |
| GenBank mRNA | AK014954.1 | - | 1 - 2 | 224 | 89 | |
| AK082932.1 | - | 1 - 2 | 92 | 89 | ||
| AK088939.1 | - | 1 - 2 | 98 | 89 | ||
| AK151235.1 | - | 1 - 2 | 98 | 89 | ||
| AK151938.1 | - | 1 - 2 | 90 | 89 | ||
| AK152679.1 | - | 1 - 2 | 90 | 89 | ||
| AK153346.1 | - | 1 - 2 | 98 | 89 | ||
| BC005621.1 | - | 1 - 2 | 78 | 3711611 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lims1
Genomic Map