Assay Details
| Gene Symbol: Mfsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114662.1 | NP_001108134.1 | 6 - 7 | 1307 | 66 | |
| NM_172510.4 | NP_766098.2 | 6 - 7 | 1307 | 66 | ||
| GenBank mRNA | AK038882.1 | - | 6 - 7 | 1200 | 66 | |
| AK044667.1 | - | 6 - 7 | 1181 | 66 | ||
| AK048692.1 | - | 5 - 6 | 1109 | 66 | ||
| AK162495.1 | - | 6 - 7 | 1211 | 66 | ||
| BC058790.1 | - | 6 - 7 | 1156 | 6395070 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd4
Genomic Map