Assay Details
| Gene Symbol: Mpzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001880.2 | NP_001001880.1 | 2 - 3 | 409 | 80 | |
| NM_001083897.1 | NP_001077366.1 | 2 - 3 | 409 | 80 | ||
| GenBank mRNA | AK076882.1 | - | 2 - 3 | 242 | 80 | |
| AK090278.1 | - | 2 - 3 | 396 | 80 | ||
| AK169385.1 | - | 2 - 3 | 399 | 80 | ||
| AK192408.1 | - | 1 - 2 | 187 | 80 | ||
| AK193736.1 | - | 2 - 3 | 187 | 80 | ||
| AK210564.1 | - | 2 - 3 | 225 | 80 | ||
| AY764247.1 | - | 2 - 3 | 406 | 80 | ||
| AY764248.1 | - | 2 - 3 | 261 | 80 | ||
| BC072563.1 | - | 2 - 3 | 404 | 6808882 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpzl1
Genomic Map