Assay Details
| Gene Symbol: R3hdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181750.2 | NP_861415.2 | 14 - 15 | 1650 | 91 | |
| GenBank mRNA | BC032160.1 | - | 5 - 6 | 541 | 91 | |
| BC049181.1 | - | 14 - 15 | 1632 | 91 | ||
| BC137772.1 | - | 13 - 14 | 1395 | 8860791 | 91 | |
| BC144899.1 | - | 13 - 14 | 1395 | 91 | ||
More Information
Set Membership: |
|

R3hdm1
Genomic Map