Assay Details
| Gene Symbol: Atp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178405.3 | NP_848492.1 | 7 - 8 | 922 | 76 | |
| GenBank mRNA | AK052863.1 | - | 7 - 8 | 859 | 76 | |
| AK129212.1 | - | 7 - 8 | 835 | 76 | ||
| AK137602.1 | - | 7 - 8 | 858 | 76 | ||
| AK147331.1 | - | 7 - 8 | 854 | 76 | ||
| AK148775.1 | - | 7 - 8 | 854 | 76 | ||
| AK161840.1 | - | 7 - 8 | 854 | 76 | ||
| AK164509.1 | - | 7 - 8 | 858 | 76 | ||
| AK168703.1 | - | 7 - 8 | 854 | 76 | ||
| BC041774.1 | - | 4 - 5 | 572 | 76 | ||
More Information
Set Membership: |
|

Atp1a2
Genomic Map