Assay Details
| Gene Symbol: Slco4c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172658.3 | NP_766246.1 | 4 - 5 | 947 | 109 | |
| GenBank mRNA | AK049253.1 | - | 4 - 5 | 947 | 109 | |
| AK049944.1 | - | 912 | 109 | |||
| AY654588.1 | - | 4 - 5 | 947 | 109 | ||
| BC119022.1 | - | 4 - 5 | 906 | 8733771 | 109 | |
More Information
Set Membership: |
|

Slco4c1
Genomic Map