Assay Details
| Gene Symbol: 5330416C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045624.1 | - | 2 - 3 | 209 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 5086 | Gm5086 |
B230311A21, EG328314 |
-158.7kb | Anti-sense | Chr. 13 - 97560000 - 97583994 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 34388 |
Gm34388 | 120.6kb | Sense | Chr. 13 - 97865811 - 97894001 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

5330416C01Rik
Genomic Map