Assay Details
| Gene Symbol: D830032E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102307.1 | 2 - 3 | 205 | 103 | ||
| GenBank mRNA | AK081398.1 | - | 113 | 103 | ||
| AK085943.1 | - | 2 - 3 | 180 | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
serine (or cysteine) peptidase inhibitor, clade B, member 8 |
Serpinb8 |
CAP-2, CAP2, NK10, Spi8, ovalbumin |
-326.2kb | Sense | Chr. 1 - 107589925 - 107610472 on NCBI Build GRCm38 |
| Downstream |
cadherin 7, type 2 |
Cdh7 |
9330156F07Rik, CDH7L1 |
2.0mb | Sense | Chr. 1 - 109982327 - 110141917 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

D830032E09Rik
Genomic Map