Assay Details
| Gene Symbol: Gm11186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046047.1 | 2 - 3 | 309 | 67 | ||
| GenBank mRNA | AK043416.1 | - | 2 - 3 | 309 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 9530068E07 gene | 9530068E07Rik |
C85061, Kct2 |
-704.0kb | Anti-sense | Chr. 11 - 52396428 - 52408723 on NCBI Build GRCm38 |
| Downstream | heat shock protein 4 | Hspa4 |
70kDa, AI317151, APG-2, Hsp110, Hsp70RY, mKIAA4025 |
140.3kb | Sense | Chr. 11 - 53259814 - 53300479 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm11186
Genomic Map