Assay Details
| Gene Symbol: 1700008C04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005755.1 | - | 2 - 3 | 264 | 107 | |
| BY705915.1 | - | 2 - 3 | 262 | 107 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 40421 |
Gm40421 | -12.2kb | Anti-sense | Chr. 12 - 50985361 - 51003783 on NCBI Build GRCm38 | |
| Downstream | G2/M-phase specific E3 ubiquitin ligase | G2e3 |
6030408C04Rik, 9030416F18, AW046379, D930034K21Rik, mKIAA1333 |
303.5kb | Sense | Chr. 12 - 51348230 - 51376986 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1700008C04Rik
Genomic Map