Assay Details
| Gene Symbol: Slamf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076693.1 | - | 143 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phosphatidylinositol glycan anchor biosynthesis, class M |
Pigm |
4933437L05Rik, AI644422, AV302080, C920011G20Rik |
-75.6kb | Sense | Chr. 1 - 172376531 - 172384099 on NCBI Build GRCm38 |
| Downstream |
immunoglobulin superfamily, member 9 |
Igsf9 |
644ETD8, Dasm1, Kiaa1355-hp, NRT1, Ncaml, mKIAA1355 |
21.6kb | Sense | Chr. 1 - 172481776 - 172498877 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Slamf9
Genomic Map