Assay Details
| Gene Symbol: Slc36a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153139.4 | NP_694779.3 | 1 - 2 | 153 | 91 | |
| GenBank mRNA | AK148943.1 | - | 1 - 2 | 83 | 91 | |
| BC138556.1 | - | 1 - 2 | 117 | 8861576 | 91 | |
| BC138558.1 | - | 1 - 2 | 117 | 8861578 | 91 | |
| CJ105223.1 | - | 154 | 91 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC138556.1, BC138558.1, NM_153139.4 transcripts.
Set Membership: |
|

Slc36a1
Genomic Map