Assay Details
| Gene Symbol: Slc36a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153139.4 | NP_694779.3 | 4 - 5 | 476 | 68 | |
| GenBank mRNA | AF453743.1 | - | 3 - 4 | 462 | 68 | |
| AY211262.1 | - | 4 - 5 | 446 | 68 | ||
| BC138556.1 | - | 4 - 5 | 440 | 8861576 | 68 | |
| BC138558.1 | - | 4 - 5 | 440 | 8861578 | 68 | |
| CJ105223.1 | - | 477 | 68 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Slc36a1
Genomic Map