| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100023029.mm.1 |
NM_021537.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023029.mm.1 |
NM_021537.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023029.mm.1 |
TC0100002836.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100023031.mm.1 |
NM_021537.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023031.mm.1 |
NM_021537.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023031.mm.1 |
TC0100002836.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100023026.mm.1 |
NM_021537.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023026.mm.1 |
NM_021537.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023026.mm.1 |
TC0100002836.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100023027.mm.1 |
NM_021537.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023027.mm.1 |
NM_021537.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023027.mm.1 |
TC0100002836.mm.1 |
MTA TC |