Assay Details
| Gene Symbol: Stk25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021537.3 | NP_067512.3 | 8 - 9 | 1155 | 59 | |
| GenBank mRNA | AK148041.1 | - | 8 - 9 | 1129 | 59 | |
| AK152917.1 | - | 8 - 9 | 1098 | 59 | ||
| BC029875.1 | - | 7 - 8 | 1873 | 59 | ||
| BC052913.1 | - | 8 - 9 | 1090 | 30057101 | 59 | |
| BC071218.1 | - | 8 - 9 | 1150 | 4012695 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stk25
Genomic Map