Assay Details
| Gene Symbol: 2810442N19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040562.1 | 1 - 2 | 364 | 70 | ||
| GenBank mRNA | AK013293.1 | - | 1 - 2 | 365 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dynamin 3 | Dnm3 |
9630020E24Rik, AW061159, B230343F03Rik, C530045C17, mKIAA0820 |
Anti-sense | Chr. 1 - 161982450 - 162478323 on NCBI Build GRCm38 | |
| Downstream |
dynamin 3, opposite strand |
Dnm3os |
6030416H16Rik, Hag2, Npn1, P/L01 |
211.8kb | Sense | Chr. 1 - 162217623 - 162225550 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

2810442N19Rik
Genomic Map