Assay Details
| Gene Symbol: Trpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019510.2 | NP_062383.2 | 6 - 7 | 1948 | 103 | |
| GenBank mRNA | AF190645.1 | - | 5 - 6 | 1538 | 103 | |
| AK032317.1 | - | 6 - 7 | 1949 | 103 | ||
| AK080619.1 | - | 6 - 7 | 1936 | 103 | ||
| BC108988.2 | - | 5 - 6 | 1537 | 103 | ||
| BC108989.2 | - | 5 - 6 | 1538 | 40047766 | 103 | |
| FJ207475.1 | - | 5 - 6 | 1538 | 103 | ||
| FJ207476.1 | - | 5 - 6 | 1538 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trpc3
Genomic Map