Assay Details
| Gene Symbol: Acbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028250.3 | NP_082526.2 | 4 - 5 | 981 | 92 | |
| GenBank mRNA | AK011781.1 | - | 4 - 5 | 974 | 92 | |
| AK167959.1 | - | 5 - 6 | 921 | 92 | ||
| BC115580.1 | - | 4 - 5 | 573 | 40102241 | 92 | |
| BC115581.1 | - | 4 - 5 | 573 | 40102242 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acbd6
Genomic Map