| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100026693.mm.1 |
NM_028749.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026693.mm.1 |
NM_028749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026693.mm.1 |
TC0100003280.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100026691.mm.1 |
NM_028749.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026691.mm.1 |
NM_028749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026691.mm.1 |
TC0100003280.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100026694.mm.1 |
NM_028749.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026694.mm.1 |
NM_028749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026694.mm.1 |
TC0100003280.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100026692.mm.1 |
NM_028749.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026692.mm.1 |
NM_028749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100026692.mm.1 |
TC0100003280.mm.1 |
MTA TC |